Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL15
Entrez Gene ID
3600
Gene Name
interleukin 15
Gene Aliases
IL-15
Chromosome Location
Chr.4: 141636596 - 141733987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000585.4 NP_000576.1 1 - 2 152 109
NM_172175.2 NP_751915.1 1 - 2 152 109
GenBank mRNA CX761071.1 - 1 - 2 162 109
U14407.1 - 1 - 2 91 109

View Full Product Details