The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190981.1 | NP_001177910.1 | 13 - 14 | 1974 | 72 |
NM_002184.3 | NP_002175.2 | 14 - 15 | 2157 | 72 | |
NM_175767.2 | NP_786943.1 | 13 - 14 | 2074 | 72 | |
NR_120480.1 | 15 - 16 | 2203 | 72 | ||
XM_017009441.1 | XP_016864930.1 | 14 - 15 | 2059 | 72 | |
XM_017009442.1 | XP_016864931.1 | 13 - 14 | 2036 | 72 | |
GenBank mRNA | AB102802.1 | - | 11 - 12 | 1661 | 72 |
BC117402.1 | - | 13 - 14 | 1956 | 72 | |
BC117404.1 | - | 13 - 14 | 1956 | 72 | |
HQ628629.1 | - | 11 - 12 | 1742 | 72 | |
HQ628630.1 | - | 11 - 12 | 1661 | 72 | |
M57230.1 | - | 14 - 15 | 2099 | 72 |