The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005565.3 | NP_005556.1 | 7 - 8 | 733 | 136 |
GenBank mRNA | AK292890.1 | - | 7 - 8 | 694 | 136 |
AK300000.1 | - | 5 - 6 | 577 | 136 | |
BC016618.1 | - | 7 - 8 | 670 | 136 | |
BT007273.1 | - | 7 - 8 | 526 | 136 | |
FN432830.1 | - | 8 - 9 | 606 | 136 | |
U20158.1 | - | 7 - 8 | 781 | 136 |