The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004621.5 | NP_004612.2 | 1 - 2 | 598 | 61 |
XM_017018221.1 | XP_016873710.1 | 1 - 2 | 688 | 61 | |
GenBank mRNA | AF080394.2 | - | 1 - 2 | 190 | 61 |
AJ006276.1 | - | 1 - 2 | 600 | 61 | |
AJ271066.1 | - | 1 - 2 | 173 | 61 | |
AJ271067.1 | - | 1 - 2 | 173 | 61 | |
AJ271068.1 | - | 1 - 2 | 173 | 61 | |
BC093658.1 | - | 1 - 2 | 237 | 61 | |
BC093660.1 | - | 1 - 2 | 237 | 61 |