The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005477.2 | NP_005468.1 | 1 - 2 | 1775 | 78 |
GenBank mRNA | AJ132429.1 | - | 1 - 2 | 1347 | 78 |
AJ238850.1 | - | 1 - 2 | 1777 | 78 |