Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAP43
Entrez Gene ID
2596
Gene Name
growth associated protein 43
Gene Aliases
B-50, PP46
Chromosome Location
Chr.3: 115623304 - 115721487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002045.3 NP_002036.1 1 - 2 414 66
GenBank mRNA AK313658.1 - 1 - 2 111 66
BC007936.2 - 1 - 2 71 66
DA701390.1 - 1 - 2 414 66
M25667.1 - 1 - 2 119 66

View Full Product Details