The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258461.1 | NP_001245390.1 | 1 - 2 | 201 | 76 |
NM_006712.4 | NP_006703.1 | 1 - 2 | 201 | 76 | |
XM_011515761.2 | XP_011514063.1 | 1 - 2 | 201 | 76 | |
XM_011515762.2 | XP_011514064.1 | 1 - 2 | 201 | 76 | |
GenBank mRNA | AB209783.1 | - | 1 - 2 | 102 | 76 |
AK223132.1 | - | 1 - 2 | 111 | 76 | |
AK292232.1 | - | 1 - 2 | 86 | 76 | |
BC000377.2 | - | 1 - 2 | 113 | 76 | |
BC011770.2 | - | 1 - 2 | 112 | 76 | |
X86779.1 | - | 1 - 2 | 105 | 76 |