The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037281.1 | NP_001032358.1 | 1 - 2 | 155 | 64 |
NM_016948.2 | NP_058644.1 | 1 - 2 | 155 | 64 | |
XM_005255977.3 | XP_005256034.1 | 1 - 2 | 147 | 64 | |
XM_011523095.2 | XP_011521397.1 | 1 - 2 | 147 | 64 | |
GenBank mRNA | AB041642.1 | - | 1 - 2 | 120 | 64 |
AB043634.1 | - | 1 - 2 | 144 | 64 | |
AF252292.1 | - | 1 - 2 | 196 | 64 | |
AF265565.1 | - | 1 - 2 | 129 | 64 | |
AJ277095.1 | - | 1 - 2 | 115 | 64 | |
BC015626.2 | - | 1 - 2 | 135 | 64 |