The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000751.2 | NP_000742.1 | 7 - 8 | 875 | 64 |
NM_001256657.1 | NP_001243586.1 | 6 - 7 | 830 | 64 | |
NM_001311196.1 | NP_001298125.1 | 7 - 8 | 843 | 64 | |
XM_011510524.2 | XP_011508826.1 | 6 - 7 | 737 | 64 | |
GenBank mRNA | AK291526.1 | - | 7 - 8 | 847 | 64 |
AK296704.1 | - | 6 - 7 | 737 | 64 | |
AK300109.1 | - | 6 - 7 | 830 | 64 | |
AK315297.1 | - | 7 - 8 | 847 | 64 | |
BC093923.1 | - | 7 - 8 | 851 | 64 | |
BC093925.1 | - | 7 - 8 | 851 | 64 | |
CD013895.1 | - | 7 - 8 | 788 | 64 | |
X55019.1 | - | 7 - 8 | 823 | 64 |