The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001552.2 | NP_001543.2 | 1 - 2 | 660 | 91 |
GenBank mRNA | AK304572.1 | - | 2 - 3 | 256 | 91 |
BC016041.1 | - | 1 - 2 | 470 | 91 | |
BT019891.1 | - | 1 - 2 | 348 | 91 | |
BT019892.1 | - | 1 - 2 | 348 | 91 | |
M38177.1 | - | 1 - 2 | 373 | 91 | |
M62403.1 | - | 1 - 2 | 431 | 91 |