The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006653.4 | NP_006644.1 | 4 - 5 | 504 | 55 |
XM_011514254.2 | XP_011512556.1 | 4 - 5 | 1357 | 55 | |
XM_011514255.2 | XP_011512557.1 | 4 - 5 | 1352 | 55 | |
XM_017010193.1 | XP_016865682.1 | 4 - 5 | 2112 | 55 | |
GenBank mRNA | AF036718.1 | - | 3 - 4 | 383 | 55 |
BC010611.1 | - | 3 - 4 | 322 | 55 | |
BM741175.1 | - | 4 - 5 | 491 | 55 | |
CR457026.1 | - | 2 - 3 | 247 | 55 |