Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIN1
Entrez Gene ID
274
Gene Name
bridging integrator 1
Gene Aliases
AMPH2, AMPHL, SH3P9
Chromosome Location
Chr.2: 127048023 - 127107400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320632.1 NP_001307561.1 2 - 3 552 69
NM_001320633.1 NP_001307562.1 2 - 3 552 69
NM_001320634.1 NP_001307563.1 2 - 3 318 69
NM_001320640.1 NP_001307569.1 2 - 3 552 69
NM_001320641.1 NP_001307570.1 2 - 3 552 69
NM_001320642.1 NP_001307571.1 2 - 3 1046 69
NM_004305.3 NP_004296.1 2 - 3 552 69
NM_139343.2 NP_647593.1 2 - 3 552 69
NM_139344.2 NP_647594.1 2 - 3 552 69
NM_139345.2 NP_647595.1 2 - 3 552 69
NM_139346.2 NP_647596.1 2 - 3 552 69
NM_139347.2 NP_647597.1 2 - 3 552 69
NM_139348.2 NP_647598.1 2 - 3 552 69
NM_139349.2 NP_647599.1 2 - 3 552 69
NM_139350.2 NP_647600.1 2 - 3 552 69
NM_139351.2 NP_647601.1 2 - 3 552 69
XM_005263642.3 XP_005263699.1 2 - 3 625 69
XM_005263643.3 XP_005263700.1 2 - 3 625 69
XM_005263645.2 XP_005263702.1 2 - 3 109 69
XM_006712425.3 XP_006712488.1 2 - 3 625 69
XM_006712426.3 XP_006712489.1 2 - 3 625 69
XM_006712427.2 XP_006712490.1 2 - 3 109 69
XM_006712428.3 XP_006712491.1 2 - 3 625 69
XM_006712429.3 XP_006712492.1 2 - 3 625 69
XM_006712431.3 XP_006712494.1 2 - 3 625 69
XM_006712432.3 XP_006712495.1 2 - 3 625 69
XM_006712433.3 XP_006712496.1 2 - 3 625 69
XM_006712434.3 XP_006712497.1 2 - 3 625 69
XM_011510975.2 XP_011509277.1 2 - 3 1258 69
XM_017003819.1 XP_016859308.1 2 - 3 439 69
XM_017003820.1 XP_016859309.1 2 - 3 625 69
XM_017003821.1 XP_016859310.1 2 - 3 625 69
XM_017003822.1 XP_016859311.1 2 - 3 625 69
XM_017003823.1 XP_016859312.1 2 - 3 109 69
XM_017003824.1 XP_016859313.1 2 - 3 109 69
XM_017003825.1 XP_016859314.1 2 - 3 439 69
XM_017003826.1 XP_016859315.1 2 - 3 109 69
XM_017003827.1 XP_016859316.1 2 - 3 625 69
XM_017003828.1 XP_016859317.1 2 - 3 625 69
GenBank mRNA AF001383.1 - 2 - 3 339 69
AF004015.1 - 2 - 3 168 69
AF043898.1 - 2 - 3 168 69
AF043899.1 - 2 - 3 168 69
AF043900.1 - 2 - 3 168 69
AF043901.1 - 2 - 3 168 69
AF068914.1 - 2 - 3 228 69
AF068915.1 - 2 - 3 228 69
AF068916.1 - 2 - 3 228 69
AF068917.1 - 2 - 3 228 69
AF068918.1 - 2 - 3 228 69
AK295300.1 - 2 - 3 270 69
AK301153.1 - 2 - 3 318 69
AL537027.3 - 2 - 3 306 69
AL713697.1 - 2 - 3 513 69
BC004101.2 - 2 - 3 228 69
BM742027.1 - 2 - 3 270 69
BT006865.1 - 2 - 3 168 69
U68485.1 - 2 - 3 214 69
U87558.1 - 2 - 3 189 69

View Full Product Details