The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292032.1 | NP_001278961.1 | 5 - 6 | 655 | 72 |
NM_003427.4 | NP_003418.2 | 5 - 6 | 655 | 72 | |
XM_011514849.2 | XP_011513151.1 | 5 - 6 | 639 | 72 | |
XM_011514850.2 | XP_011513152.1 | 5 - 6 | 767 | 72 | |
XM_017011249.1 | XP_016866738.1 | 5 - 6 | 767 | 72 | |
XM_017011253.1 | XP_016866742.1 | 5 - 6 | 767 | 72 | |
XM_017011254.1 | XP_016866743.1 | 5 - 6 | 639 | 72 | |
XM_017011256.1 | XP_016866745.1 | 5 - 6 | 782 | 72 | |
GenBank mRNA | AK294605.1 | - | 5 - 6 | 639 | 72 |
AK295276.1 | - | 5 - 6 | 757 | 72 | |
AK302903.1 | - | 5 - 6 | 757 | 72 | |
BC000133.2 | - | 5 - 6 | 635 | 72 | |
BC002549.2 | - | 5 - 6 | 636 | 72 | |
BT007316.1 | - | 4 - 5 | 431 | 72 | |
M91592.1 | - | 5 - 6 | 594 | 72 |