Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3G
Entrez Gene ID
8666
Gene Name
eukaryotic translation initiation factor 3 subunit G
Gene Aliases
EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44
Chromosome Location
Chr.19: 10115014 - 10119923 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF020833.1 - 5 - 6 326 124
AF094850.1 - 5 - 6 344 124
AK291320.1 - 5 - 6 345 124
AK293374.1 - 5 - 6 429 124
AK316040.1 - 4 - 5 474 124
AK316335.1 - 4 - 5 480 124
BC000733.2 - 5 - 6 345 124
BC008469.1 - 5 - 6 346 124
BT006889.1 - 5 - 6 303 124
CR457134.1 - 5 - 6 303 124
DC356593.1 - 5 - 6 368 124
U96074.1 - 5 - 6 312 124

View Full Product Details