The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198616.1 | NP_001185545.1 | 1 - 2 | 481 | 120 |
NM_001198617.1 | NP_001185546.1 | 1 - 2 | 481 | 120 | |
NM_003980.4 | NP_003971.1 | 1 - 2 | 481 | 120 | |
XM_006715601.3 | XP_006715664.1 | 1 - 2 | 677 | 120 | |
XM_011536245.1 | XP_011534547.1 | 1 - 2 | 175 | 120 | |
GenBank mRNA | AK312789.1 | - | 1 - 2 | 81 | 120 |
AL042021.1 | - | 1 - 2 | 481 | 120 | |
BC025777.1 | - | 1 - 2 | 77 | 120 | |
CD557798.1 | - | 1 - 2 | 321 | 120 | |
DA329576.1 | - | 1 - 2 | 292 | 120 | |
X73882.1 | - | 1 - 2 | 170 | 120 |