The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004204.3 | NP_004195.2 | 7 - 8 | 1471 | 66 |
NM_148920.2 | NP_683721.1 | 7 - 8 | 1471 | 66 | |
GenBank mRNA | AB003723.1 | - | 7 - 8 | 1435 | 66 |
AF030177.1 | - | 7 - 8 | 1404 | 66 | |
AK055163.1 | - | 1 - 2 | 122 | 66 | |
AK300297.1 | - | 7 - 8 | 1438 | 66 | |
AK314361.1 | - | 8 - 9 | 1500 | 66 | |
BC006377.2 | - | 7 - 8 | 1421 | 66 |