The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204051.1 | NP_001190980.1 | 3 - 4 | 664 | 61 |
NM_001204052.1 | NP_001190981.1 | 3 - 4 | 664 | 61 | |
NM_004277.4 | NP_004268.3 | 3 - 4 | 664 | 61 | |
XM_005249483.2 | XP_005249540.1 | 3 - 4 | 634 | 61 | |
XM_017011512.1 | XP_016867001.1 | 2 - 3 | 513 | 61 | |
GenBank mRNA | AF110532.1 | - | 3 - 4 | 386 | 61 |
AK090871.1 | - | 1 - 2 | 993 | 61 | |
AK291427.1 | - | 3 - 4 | 594 | 61 | |
AK295233.1 | - | 3 - 4 | 647 | 61 | |
AK302906.1 | - | 4 - 5 | 704 | 61 | |
AK315940.1 | - | 3 - 4 | 575 | 61 | |
AY358711.1 | - | 3 - 4 | 412 | 61 | |
BC033091.1 | - | 3 - 4 | 637 | 61 | |
BP310559.1 | - | 4 - 5 | 551 | 61 | |
DR002070.1 | - | 3 - 4 | 586 | 61 |