The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165928.3 | NP_001159400.2 | 5 - 6 | 1066 | 96 |
NM_001177634.2 | NP_001171105.1 | 5 - 6 | 1009 | 96 | |
NM_001177635.2 | NP_001171106.1 | 3 - 4 | 598 | 96 | |
NM_001177636.2 | NP_001171107.1 | 4 - 5 | 987 | 96 | |
NM_001177637.2 | NP_001171108.1 | 3 - 4 | 825 | 96 | |
NM_001177638.2 | NP_001171109.1 | 3 - 4 | 856 | 96 | |
NM_001177639.2 | NP_001171110.1 | 2 - 3 | 546 | 96 | |
NM_001177640.2 | NP_001171111.1 | 3 - 4 | 762 | 96 | |
NM_001177641.2 | NP_001171112.1 | 3 - 4 | 852 | 96 | |
NM_001177642.2 | NP_001171113.1 | 3 - 4 | 757 | 96 | |
NM_001177643.2 | NP_001171114.1 | 2 - 3 | 756 | 96 | |
NM_001177644.2 | NP_001171115.1 | 2 - 3 | 845 | 96 | |
NM_004393.5 | NP_004384.4 | 2 - 3 | 705 | 96 | |
GenBank mRNA | AK291692.1 | - | 2 - 3 | 547 | 96 |
BC012740.2 | - | 2 - 3 | 640 | 96 | |
BC014616.2 | - | 2 - 3 | 640 | 96 | |
DA223570.1 | - | 3 - 4 | 483 | 96 | |
L19711.1 | - | 2 - 3 | 681 | 96 |