Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISG15
Entrez Gene ID
9636
Gene Name
ISG15 ubiquitin-like modifier
Gene Aliases
G1P2, IFI15, IMD38, IP17, UCRP, hUCRP
Chromosome Location
Chr.1: 1013467 - 1014540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005101.3 NP_005092.1 1 - 2 115 100
GenBank mRNA BM712238.1 - 1 - 2 116 100
BU161226.1 - 1 - 2 72 100
M13755.1 - 1 - 2 83 100

View Full Product Details