The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005478.4 | NP_005469.2 | 1 - 2 | 218 | 119 |
GenBank mRNA | AF133816.2 | - | 1 - 2 | 227 | 119 |
AY359030.1 | - | 1 - 2 | 218 | 119 | |
BC101646.1 | - | 1 - 2 | 207 | 119 | |
BC101648.1 | - | 1 - 2 | 207 | 119 |