The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282206.1 | NP_001269135.1 | 3 - 4 | 525 | 79 |
NM_001282207.1 | NP_001269136.1 | 3 - 4 | 512 | 79 | |
NM_001282208.1 | NP_001269137.1 | 3 - 4 | 595 | 79 | |
NM_005866.3 | NP_005857.1 | 3 - 4 | 572 | 79 | |
NR_104108.1 | 3 - 4 | 459 | 79 | ||
GenBank mRNA | AK098451.1 | - | 4 - 5 | 363 | 79 |
AK130502.1 | - | 3 - 4 | 555 | 79 | |
AK222899.1 | - | 3 - 4 | 501 | 79 | |
AY633611.1 | - | 3 - 4 | 507 | 79 | |
BC004899.2 | - | 3 - 4 | 478 | 79 | |
BC007839.2 | - | 3 - 4 | 442 | 79 | |
BI596428.1 | - | 3 - 4 | 551 | 79 | |
CR457075.1 | - | 3 - 4 | 443 | 79 | |
DQ644568.1 | - | 3 - 4 | 383 | 79 | |
DQ647702.1 | - | 3 - 4 | 396 | 79 | |
U75283.1 | - | 3 - 4 | 490 | 79 | |
U79528.1 | - | 3 - 4 | 517 | 79 |