The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005909.4 | NP_005900.2 | 2 - 3 | 550 | 88 |
GenBank mRNA | AK308147.1 | - | 2 - 3 | 341 | 88 |
BC017240.2 | - | 2 - 3 | 527 | 88 | |
BC025240.1 | - | 2 - 3 | 469 | 88 | |
BC032516.1 | - | 2 - 3 | 342 | 88 | |
BC039822.1 | - | 2 - 3 | 336 | 88 | |
BC046114.1 | - | 2 - 3 | 356 | 88 | |
BC056145.1 | - | 2 - 3 | 330 | 88 | |
BC063669.1 | - | 2 - 3 | 330 | 88 | |
BC073993.1 | - | 2 - 3 | 332 | 88 | |
BC094834.1 | - | 2 - 3 | 327 | 88 | |
BC108733.1 | - | 2 - 3 | 345 | 88 | |
BC139918.1 | - | 2 - 3 | 447 | 88 | |
BC150196.1 | - | 2 - 3 | 447 | 88 | |
BX647077.1 | - | 2 - 3 | 341 | 88 |