Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRDN
Entrez Gene ID
10345
Gene Name
triadin
Gene Aliases
CPVT5, TDN, TRISK
Chromosome Location
Chr.6: 123216339 - 123637093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251987.1 NP_001238916.1 13 - 14 1427 74
NM_006073.3 NP_006064.2 13 - 14 1424 74
GenBank mRNA AJ489257.2 - 13 - 14 1322 74
CR749591.1 - 3 - 4 183 74
U18985.1 - 13 - 14 1128 74

View Full Product Details