The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143835.1 | NP_001137307.1 | 16 - 17 | 1773 | 126 |
NM_006165.3 | NP_006156.2 | 14 - 15 | 1768 | 126 | |
XM_005271567.3 | XP_005271624.1 | 16 - 17 | 1809 | 126 | |
XM_005271568.3 | XP_005271625.1 | 15 - 16 | 1724 | 126 | |
XM_011542850.2 | XP_011541152.1 | 17 - 18 | 1846 | 126 | |
XM_011542851.2 | XP_011541153.1 | 16 - 17 | 1828 | 126 | |
XM_011542852.2 | XP_011541154.1 | 15 - 16 | 1666 | 126 | |
XM_017017793.1 | XP_016873282.1 | 15 - 16 | 1720 | 126 | |
XM_017017794.1 | XP_016873283.1 | 15 - 16 | 1684 | 126 | |
XM_017017795.1 | XP_016873284.1 | 17 - 18 | 1810 | 126 | |
XM_017017796.1 | XP_016873285.1 | 15 - 16 | 1688 | 126 | |
XM_017017797.1 | XP_016873286.1 | 15 - 16 | 1720 | 126 | |
XM_017017798.1 | XP_016873287.1 | 16 - 17 | 1773 | 126 | |
XM_017017799.1 | XP_016873288.1 | 17 - 18 | 1920 | 126 | |
XM_017017800.1 | XP_016873289.1 | 17 - 18 | 1860 | 126 | |
GenBank mRNA | BC063280.1 | - | 16 - 17 | 1719 | 126 |
X80878.1 | - | 14 - 15 | 1777 | 126 |