The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006417.4 | NP_006408.3 | 2 - 3 | 587 | 107 |
NR_135641.1 | 2 - 3 | 590 | 107 | ||
XM_005270380.3 | XP_005270437.1 | 2 - 3 | 1825 | 107 | |
XM_011540516.2 | XP_011538818.1 | 2 - 3 | 1825 | 107 | |
XM_017000083.1 | XP_016855572.1 | 2 - 3 | 1825 | 107 | |
GenBank mRNA | AK302527.1 | - | 2 - 3 | 548 | 107 |
AK316476.1 | - | 2 - 3 | 545 | 107 | |
BC022870.1 | - | 2 - 3 | 552 | 107 | |
DN994910.1 | - | 2 - 3 | 564 | 107 |