The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286474.1 | NP_001273403.1 | 20 - 21 | 736 | 112 |
NM_001286475.1 | NP_001273404.1 | 19 - 20 | 715 | 91 | |
XM_017010182.1 | XP_016865671.1 | 9 - 10 | 206 | 169 | |
XM_017010183.1 | XP_016865672.1 | 9 - 10 | 745 | 112 | |
XM_017010184.1 | XP_016865673.1 | 8 - 9 | 724 | 112 | |
GenBank mRNA | AK302176.1 | - | 19 - 20 | 675 | 91 |
AK310230.1 | - | 19 - 20 | 704 | 112 | |
U60665.1 | - | 21 - 22 | 784 | 91 |