The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300791.1 | NP_001287720.1 | 5 - 6 | 779 | 105 |
NM_001300792.1 | NP_001287721.1 | 5 - 6 | 779 | 105 | |
NM_007054.6 | NP_008985.3 | 5 - 6 | 779 | 105 | |
XM_006714526.3 | XP_006714589.1 | 5 - 6 | 834 | 105 | |
XM_017008995.1 | XP_016864484.1 | 5 - 6 | 834 | 105 | |
XM_017008996.1 | XP_016864485.1 | 5 - 6 | 834 | 105 | |
GenBank mRNA | AB209780.1 | - | 5 - 6 | 727 | 105 |
AF041853.1 | - | 4 - 5 | 620 | 105 | |
AK295089.1 | - | 5 - 6 | 737 | 105 | |
AK313359.1 | - | 5 - 6 | 747 | 105 | |
AM177178.1 | - | 4 - 5 | 620 | 105 | |
BC020890.1 | - | 5 - 6 | 744 | 105 | |
BC032599.1 | - | 5 - 6 | 762 | 105 | |
BC039592.1 | - | 5 - 6 | 736 | 105 | |
BC045542.1 | - | 5 - 6 | 773 | 105 |