The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281428.1 | NP_001268357.1 | 10 - 11 | 1167 | 72 |
NM_012144.3 | NP_036276.1 | 10 - 11 | 1155 | 72 | |
XM_006716758.3 | XP_006716821.1 | 9 - 10 | 798 | 72 | |
XM_011517846.1 | XP_011516148.1 | 10 - 11 | 1167 | 72 | |
XM_011517847.2 | XP_011516149.1 | 10 - 11 | 1167 | 72 | |
XM_011517848.1 | XP_011516150.1 | 10 - 11 | 1167 | 72 | |
XM_011517849.1 | XP_011516151.1 | 10 - 11 | 1167 | 72 | |
XM_011517850.2 | XP_011516152.1 | 10 - 11 | 1167 | 72 | |
XM_017014625.1 | XP_016870114.1 | 10 - 11 | 1155 | 72 | |
GenBank mRNA | AF091619.1 | - | 10 - 11 | 1072 | 72 |
AK302186.1 | - | 7 - 8 | 698 | 72 | |
AK303937.1 | - | 7 - 8 | 654 | 72 | |
AK314639.1 | - | 10 - 11 | 1067 | 72 | |
BC030583.2 | - | 10 - 11 | 1083 | 72 |