The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013323.2 | NP_037455.2 | 4 - 5 | 500 | 87 |
NM_152244.1 | NP_689450.1 | 5 - 6 | 587 | 87 | |
XM_005257260.3 | XP_005257317.1 | 4 - 5 | 267 | 87 | |
XM_005257261.3 | XP_005257318.1 | 4 - 5 | 534 | 87 | |
XM_005257262.3 | XP_005257319.1 | 4 - 5 | 328 | 87 | |
XM_011524697.2 | XP_011522999.1 | 5 - 6 | 370 | 87 | |
GenBank mRNA | AF121861.1 | - | 4 - 5 | 520 | 87 |
AK023932.1 | - | 5 - 6 | 587 | 87 | |
AK091852.1 | - | 5 - 6 | 485 | 87 | |
AK296095.1 | - | 2 - 3 | 394 | 87 | |
AK298551.1 | - | 4 - 5 | 323 | 87 | |
AK316374.1 | - | 4 - 5 | 320 | 87 | |
BC000768.2 | - | 4 - 5 | 317 | 87 | |
BC103721.1 | - | 4 - 5 | 306 | 87 | |
BT006723.1 | - | 3 - 4 | 233 | 87 |