The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013363.3 | NP_037495.1 | 3 - 4 | 758 | 71 |
GenBank mRNA | AF098269.1 | - | 3 - 4 | 484 | 71 |
AK075478.1 | - | 3 - 4 | 755 | 71 | |
AK223633.1 | - | 3 - 4 | 680 | 71 | |
AK315121.1 | - | 3 - 4 | 526 | 71 | |
AY035400.1 | - | 3 - 4 | 647 | 71 | |
AY358557.1 | - | 3 - 4 | 757 | 71 | |
BC006265.2 | - | 3 - 4 | 511 | 71 | |
CR536570.1 | - | 3 - 4 | 451 | 71 |