Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGA1
Entrez Gene ID
26088
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
-
Chromosome Location
Chr.22: 37607235 - 37633564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001560.2 NP_001001560.1 1 - 2 430 71
NM_001172687.1 NP_001166158.1 1 - 2 430 71
NM_013365.4 NP_037497.1 1 - 2 430 71
XM_005261517.3 XP_005261574.1 2 - 3 604 71
XM_011530122.1 XP_011528424.1 2 - 3 537 71
XM_011530124.1 XP_011528426.1 2 - 3 604 71
XM_017028760.1 XP_016884249.1 2 - 3 604 71
XM_017028762.1 XP_016884251.1 2 - 3 604 71
XM_017028763.1 XP_016884252.1 1 - 2 70 71
GenBank mRNA AF190862.1 - 1 - 2 58 71
AF218584.1 - 1 - 2 47 71
AF233521.1 - 1 - 2 65 71
AK075256.1 - 1 - 2 408 71
AK225157.1 - 1 - 2 64 71
AK290548.1 - 1 - 2 69 71
BC010917.1 - 1 - 2 74 71
BC029388.1 - 1 - 2 69 71
BC044629.1 - 1 - 2 59 71
CR456493.1 - 1 - 2 179 71

View Full Product Details