Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ST6GALNAC6
- Entrez Gene ID
-
30815
- Gene Name
- ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
- Gene Aliases
-
SIAT7-F, SIAT7F, ST6GALNACVI
- Chromosome Location
-
Chr.9: 127885321 - 127906725 on Build GRCh38
View Full Product Details