The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014176.3 | NP_054895.1 | 2 - 3 | 242 | 115 |
GenBank mRNA | AB032931.1 | - | 1 - 2 | 148 | 115 |
AF160215.1 | - | 2 - 3 | 199 | 115 | |
AF161499.1 | - | 2 - 3 | 232 | 115 | |
AK000504.1 | - | 2 - 3 | 197 | 115 | |
AY542309.1 | - | 1 - 2 | 107 | 115 | |
BC004152.2 | - | 2 - 3 | 204 | 115 | |
BC019284.1 | - | 2 - 3 | 204 | 115 | |
BG776416.1 | - | 2 - 3 | 243 | 115 | |
BP244265.1 | - | 2 - 3 | 242 | 115 |