Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
Chromosome Location
Chr.8: 39584568 - 39730064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320313.1 NP_001307242.1 13 - 14 1508 137
NM_014237.2 NP_055052.1 14 - 15 1580 137
NR_135201.1 12 - 13 1218 137
GenBank mRNA AJ133004.1 - 14 - 15 1525 137
AK313961.1 - 14 - 15 1570 137
AY358321.1 - 13 - 14 1495 137
BC121045.1 - 14 - 15 1525 137
BC121046.1 - 12 - 13 1163 137

View Full Product Details