The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014462.2 | NP_055277.1 | 3 - 4 | 640 | 120 |
NR_045493.1 | 3 - 4 | 362 | 120 | ||
GenBank mRNA | AF000177.1 | - | 3 - 4 | 394 | 120 |
AF147348.1 | - | 3 - 4 | 283 | 120 | |
AJ238094.1 | - | 3 - 4 | 418 | 120 | |
AK312159.1 | - | 3 - 4 | 393 | 120 | |
BC001767.1 | - | 3 - 4 | 327 | 120 | |
BE670269.1 | - | 1 - 2 | 120 | ||
BU941962.1 | - | 3 - 4 | 365 | 120 | |
KU178606.1 | - | 3 - 4 | 230 | 120 |