The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009568.2 | NP_001009568.1 | 1 - 2 | 289 | 75 |
NM_001304579.1 | NP_001291508.1 | 1 - 2 | 289 | 75 | |
NM_014474.3 | NP_055289.2 | 1 - 2 | 289 | 75 | |
GenBank mRNA | AK293489.1 | - | 1 - 2 | 135 | 75 |
AK293566.1 | - | 1 - 2 | 215 | 75 | |
AK301349.1 | - | 1 - 2 | 244 | 75 | |
AK316500.1 | - | 1 - 2 | 215 | 75 | |
BC014444.1 | - | 1 - 2 | 206 | 75 | |
BP284513.1 | - | 1 - 2 | 290 | 75 | |
CB160850.1 | - | 1 - 2 | 251 | 75 | |
DC370191.1 | - | 1 - 2 | 288 | 75 | |
Y08134.1 | - | 1 - 2 | 181 | 75 |