The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195555.1 | NP_001182484.1 | 6 - 7 | 900 | 103 |
NM_001195556.1 | NP_001182485.1 | 6 - 7 | 877 | 103 | |
NM_014666.3 | NP_055481.1 | 6 - 7 | 900 | 103 | |
XM_011534701.2 | XP_011533003.1 | 6 - 7 | 885 | 103 | |
XM_017010087.1 | XP_016865576.1 | 6 - 7 | 885 | 103 | |
XM_017010088.1 | XP_016865577.1 | 6 - 7 | 885 | 103 | |
GenBank mRNA | AF434813.1 | - | 6 - 7 | 695 | 103 |
AK092765.1 | - | 6 - 7 | 847 | 103 | |
AK300257.1 | - | 6 - 7 | 868 | 103 | |
BC004467.2 | - | 6 - 7 | 761 | 103 | |
D79993.1 | - | 6 - 7 | 796 | 103 | |
KU178486.1 | - | 6 - 7 | 695 | 103 | |
KU178487.1 | - | 6 - 7 | 695 | 103 |