The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014788.3 | NP_055603.2 | 3 - 4 | 703 | 125 |
NM_033219.2 | NP_150088.1 | 3 - 4 | 703 | 125 | |
XM_005252320.3 | XP_005252377.1 | 3 - 4 | 768 | 125 | |
XM_006717338.1 | XP_006717401.1 | 3 - 4 | 1326 | 125 | |
XM_011519244.2 | XP_011517546.1 | 3 - 4 | 533 | 125 | |
XM_017015352.1 | XP_016870841.1 | 3 - 4 | 768 | 125 | |
XM_017015353.1 | XP_016870842.1 | 3 - 4 | 768 | 125 | |
XM_017015354.1 | XP_016870843.1 | 3 - 4 | 759 | 125 | |
GenBank mRNA | AF220130.1 | - | 3 - 4 | 548 | 125 |
AF220131.1 | - | 3 - 4 | 548 | 125 | |
AK055833.1 | - | 3 - 4 | 576 | 125 | |
AK097480.1 | - | 3 - 4 | 547 | 125 | |
AK292825.1 | - | 3 - 4 | 570 | 125 | |
BC006333.2 | - | 3 - 4 | 556 | 125 | |
D50919.1 | - | 3 - 4 | 548 | 125 |