The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014840.2 | NP_055655.1 | 1 - 2 | 1624 | 66 |
GenBank mRNA | AB011109.1 | - | 1 - 2 | 1624 | 66 |
AK289992.1 | - | 1 - 2 | 304 | 66 | |
BC152462.1 | - | 1 - 2 | 1624 | 66 |