The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014879.3 | NP_055694.3 | 1 - 2 | 153 | 85 |
GenBank mRNA | BC034989.2 | - | 1 - 2 | 77 | 85 |
D13626.1 | - | 1 - 2 | 81 | 85 |