Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMMECR1
Entrez Gene ID
9949
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
AMMERC1
Chromosome Location
Chr.X: 110194186 - 110440233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171689.1 NP_001165160.1 3 - 4 743 105
NM_015365.2 NP_056180.1 1 - 2 556 105
GenBank mRNA AJ007014.1 - 1 - 2 643 105
AK001703.1 - 1 - 2 79 105
AK091430.1 - 3 - 4 743 105
AK314596.1 - 1 - 2 487 105
KU178497.1 - 1 - 2 475 105

View Full Product Details