The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015836.3 | NP_056651.1 | 3 - 4 | 454 | 109 |
NM_201263.2 | NP_957715.1 | 3 - 4 | 454 | 109 | |
XM_005270350.3 | XP_005270407.1 | 3 - 4 | 540 | 109 | |
XM_011540494.2 | XP_011538796.1 | 4 - 5 | 523 | 109 | |
XM_011540495.2 | XP_011538797.1 | 2 - 3 | 225 | 109 | |
XM_017000039.1 | XP_016855528.1 | 5 - 6 | 709 | 109 | |
XM_017000040.1 | XP_016855529.1 | 3 - 4 | 276 | 109 | |
GenBank mRNA | AJ242739.1 | - | 3 - 4 | 435 | 109 |
AK223197.1 | - | 3 - 4 | 432 | 109 | |
AK299547.1 | - | 3 - 4 | 393 | 109 | |
AK300295.1 | - | 3 - 4 | 282 | 109 | |
AK313740.1 | - | 3 - 4 | 452 | 109 | |
BC021722.1 | - | 3 - 4 | 433 | 109 | |
BC039889.1 | - | 3 - 4 | 430 | 109 | |
BC044575.1 | - | 3 - 4 | 437 | 109 | |
BM834499.1 | - | 3 - 4 | 446 | 109 |