The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016538.2 | NP_057622.1 | 3 - 4 | 384 | 77 |
GenBank mRNA | AF233395.1 | - | 3 - 4 | 366 | 77 |
AK002027.1 | - | 3 - 4 | 354 | 77 | |
AK094326.1 | - | 2 - 3 | 449 | 77 | |
AK290265.1 | - | 3 - 4 | 352 | 77 | |
BC017305.2 | - | 3 - 4 | 348 | 77 | |
BC101791.1 | - | 3 - 4 | 365 | 77 | |
BC101793.1 | - | 3 - 4 | 364 | 77 | |
BQ890203.1 | - | 3 - 4 | 384 | 77 | |
CD579273.2 | - | 3 - 4 | 370 | 77 |