The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202560.2 | NP_001189489.1 | 5 - 6 | 422 | 80 |
NM_017528.4 | NP_059998.2 | 5 - 6 | 422 | 80 | |
NR_037776.2 | 5 - 6 | 618 | 80 | ||
XM_006715847.1 | XP_006715910.1 | 5 - 6 | 515 | 80 | |
XM_011515778.1 | XP_011514080.1 | 5 - 6 | 515 | 80 | |
XM_011515779.1 | XP_011514081.1 | 5 - 6 | 419 | 80 | |
GenBank mRNA | AF218007.1 | - | 4 - 5 | 314 | 80 |
AF412034.1 | - | 5 - 6 | 422 | 80 | |
AF420248.1 | - | 5 - 6 | 400 | 80 | |
AJ224442.1 | - | 5 - 6 | 381 | 80 | |
AK291116.1 | - | 5 - 6 | 393 | 80 | |
AK315032.1 | - | 5 - 6 | 419 | 80 | |
BC000169.2 | - | 5 - 6 | 378 | 80 | |
BC001780.1 | - | 5 - 6 | 401 | 80 | |
BC011696.2 | - | 5 - 6 | 383 | 80 | |
BQ927478.1 | - | - | 363 | 80 | |
BQ961799.1 | - | 5 - 6 | 586 | 80 | |
DT220127.1 | - | 5 - 6 | 422 | 80 |