The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172105.1 | NP_001165576.1 | 6 - 7 | 1009 | 107 |
NM_017577.4 | NP_060047.3 | 11 - 12 | 1282 | 107 | |
XM_005247546.2 | XP_005247603.1 | 10 - 11 | 1198 | 107 | |
XM_005247547.1 | XP_005247604.1 | 11 - 12 | 1244 | 107 | |
XM_011512930.1 | XP_011511232.1 | 11 - 12 | 1277 | 107 | |
XM_011512931.1 | XP_011511233.1 | 6 - 7 | 1066 | 107 | |
XM_017006646.1 | XP_016862135.1 | 10 - 11 | 1196 | 107 | |
XM_017006647.1 | XP_016862136.1 | 12 - 13 | 1360 | 107 | |
XM_017006648.1 | XP_016862137.1 | 7 - 8 | 1089 | 107 | |
XM_017006649.1 | XP_016862138.1 | 5 - 6 | 840 | 107 | |
GenBank mRNA | AK093181.1 | - | 6 - 7 | 801 | 107 |
AK097783.1 | - | 12 - 13 | 1366 | 107 | |
AK292846.1 | - | 11 - 12 | 1218 | 107 | |
AK292964.1 | - | 6 - 7 | 924 | 107 | |
AK293884.1 | - | 9 - 10 | 994 | 107 | |
AK304142.1 | - | 6 - 7 | 1009 | 107 | |
AL133661.1 | - | 2 - 3 | 1498 | 107 | |
AY358725.1 | - | 6 - 7 | 962 | 107 | |
BC028972.2 | - | 3 - 4 | 286 | 107 | |
BC035040.1 | - | 11 - 12 | 1288 | 107 |