Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- VPS13B
- Entrez Gene ID
-
157680
- Gene Name
- vacuolar protein sorting 13 homolog B
- Gene Aliases
-
CHS1, COH1
- Chromosome Location
-
Chr.8: 99007014 - 99877586 on Build GRCh38
View Full Product Details