The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017925.6 | NP_060395.5 | 27 - 28 | 5100 | 100 |
NR_073201.3 | 27 - 28 | 4966 | 100 | ||
XM_006716804.3 | XP_006716867.1 | 28 - 29 | 5215 | 100 | |
XM_006716805.3 | XP_006716868.1 | 28 - 29 | 5215 | 100 | |
XM_006716807.3 | XP_006716870.1 | 19 - 20 | 3740 | 100 | |
XM_017014917.1 | XP_016870406.1 | 27 - 28 | 5008 | 100 | |
XM_017014918.1 | XP_016870407.1 | 29 - 30 | 5415 | 100 | |
XM_017014919.1 | XP_016870408.1 | 29 - 30 | 5382 | 100 | |
XM_017014920.1 | XP_016870409.1 | 28 - 29 | 5239 | 100 | |
GenBank mRNA | AK000693.1 | - | 9 - 10 | 1303 | 100 |
AK001046.1 | - | 4 - 5 | 681 | 100 | |
AK002020.1 | - | 2 - 3 | 265 | 100 | |
AK025705.1 | - | 7 - 8 | 2121 | 100 | |
AK125842.1 | - | 21 - 22 | 3962 | 100 | |
AK127734.1 | - | 16 - 17 | 100 | ||
AK299842.1 | - | 14 - 15 | 3021 | 100 | |
AK302901.1 | - | 9 - 10 | 2432 | 100 | |
AL834521.1 | - | 6 - 7 | 1375 | 100 | |
BC144484.1 | - | 24 - 25 | 4822 | 100 | |
BC144485.1 | - | 23 - 24 | 4198 | 100 |