Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FEZF2
- Entrez Gene ID
-
55079
- Gene Name
- FEZ family zinc finger 2
- Gene Aliases
-
FEZ, FEZL, FKSG36, TOF, ZFP312, ZNF312
- Chromosome Location
-
Chr.3: 62369672 - 62373515 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001004.1 |
- |
2 - 3 |
1097 |
88 |
View Full Product Details