Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDYHV1
Entrez Gene ID
55093
Gene Name
WDYHV motif containing 1
Gene Aliases
C8orf32
Chromosome Location
Chr.8: 123416725 - 123467527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283024.1 NP_001269953.1 3 - 4 286 64
NM_018024.2 NP_060494.1 3 - 4 362 64
NR_133926.1 3 - 4 286 64
XM_006716597.3 XP_006716660.1 3 - 4 356 64
XM_011517147.1 XP_011515449.1 3 - 4 355 64
XM_017013601.1 XP_016869090.1 3 - 4 356 64
XM_017013602.1 XP_016869091.1 3 - 4 279 64
XM_017013603.1 XP_016869092.1 3 - 4 278 64
GenBank mRNA AK001066.1 - 3 - 4 362 64
AK225643.1 - 3 - 4 313 64
AK293492.1 - 3 - 4 206 64
AL528914.3 - 3 - 4 278 64
AU119047.1 - 3 - 4 362 64
BC008781.2 - 3 - 4 280 64

View Full Product Details