Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP7D1
Entrez Gene ID
55700
Gene Name
MAP7 domain containing 1
Gene Aliases
PARCC1, RPRC1
Chromosome Location
Chr.1: 36155965 - 36180849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286365.1 NP_001273294.1 11 - 12 2478 92
NM_001286366.1 NP_001273295.1 13 - 14 2493 92
XM_006710759.1 XP_006710822.1 12 - 13 2404 92
XM_006710761.1 XP_006710824.1 12 - 13 2197 92
GenBank mRNA AB033013.2 - 11 - 12 2478 92
AF218002.1 - 12 - 13 1815 92
AK001212.1 - 4 - 5 672 92
AK095939.1 - 5 - 6 836 92
AK096341.1 - 13 - 14 2260 92
AK309966.1 - 10 - 11 1484 92
AL136547.1 - 4 - 5 503 92
BC003083.2 - 4 - 5 739 92
BC027334.1 - 2 - 3 264 92
BC057293.1 - 11 - 12 2280 92
BC067256.1 - 9 - 10 1651 92
BC082266.1 - 6 - 7 1014 92
BC128042.1 - 2 - 3 110 92
CR457254.1 - 3 - 4 369 92

View Full Product Details